Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002800.2 | NP_001002800.1 | 6 | 1183 | ||
NM_001288753.1 | NP_001275682.1 | 6 | 1108 | |||
NM_005496.3 | NP_005487.3 | 5 | 1000 | |||
XM_006713459.3 | XP_006713522.1 | 6 | 953 | |||
XM_011512311.2 | XP_011510613.1 | 6 | 1071 | |||
XM_011512312.2 | XP_011510614.1 | 5 | 692 | |||
XM_017005491.1 | XP_016860980.1 | 4 | 518 | |||
GenBank mRNA | AB019987.1 | - | 6 | 844 | ||
AF092564.1 | - | 4 | 474 | |||
AK002200.1 | - | 6 | 837 | |||
AK128143.1 | - | 6 | 762 | |||
AK225437.1 | - | 7 | 837 | |||
AK307508.1 | - | 299 | ||||
AK315070.1 | - | 6 | 837 | |||
AL136877.1 | - | 5 | 964 | |||
AL833949.1 | - | 5 | 1891 | |||
BC001557.1 | - | 6 | 819 | |||
BC065259.1 | - | 6 | 1015 | |||
BC070161.1 | - | 6 | 837 | |||
BC106033.1 | - | 6 | 855 |