Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002800.2 | NP_001002800.1 | 18 | 3192 | ||
NM_001288753.1 | NP_001275682.1 | 18 | 3117 | |||
NM_005496.3 | NP_005487.3 | 17 | 3009 | |||
XM_006713459.3 | XP_006713522.1 | 17 | 2728 | |||
XM_011512311.2 | XP_011510613.1 | 18 | 3080 | |||
XM_011512312.2 | XP_011510614.1 | 17 | 2701 | |||
XM_017005491.1 | XP_016860980.1 | 16 | 2923 | |||
GenBank mRNA | AB019987.1 | - | 18 | 2853 | ||
AF092564.1 | - | 16 | 2483 | |||
AK122939.1 | - | 10 | 1965 | |||
AK128143.1 | - | 18 | 2771 | |||
AK315070.1 | - | 18 | 2848 | |||
AL136877.1 | - | 17 | 2973 | |||
AL833949.1 | - | 17 | 3900 |