Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201536.1 | NP_001188465.1 | 4 | 520 | ||
NM_005681.3 | NP_005672.1 | 4 | 520 | |||
NM_139352.2 | NP_647603.1 | 3 | 350 | |||
XM_005273343.3 | XP_005273400.1 | 4 | 683 | |||
XM_006711613.3 | XP_006711676.1 | 4 | 533 | |||
XM_017002759.1 | XP_016858248.1 | 4 | 489 | |||
XM_017002760.1 | XP_016858249.1 | 3 | 297 | |||
GenBank mRNA | AK001054.1 | - | 3 | 202 | ||
AK299536.1 | - | 4 | 520 | |||
AK315740.1 | - | 4 | 504 | |||
BC013808.1 | - | 4 | 500 | 3918866 | ||
L39060.1 | - | 3 | 335 |