Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADGRG1 SIRNA Assays ›
Gene Symbol:
ADGRG1
Gene Name:
adhesion G protein-coupled receptor G1
Gene Aliases:
BFPP, BPPR, GPR56, TM7LN4, TM7XN1
Chromosome Location:
Chr.16: 57619535 - 57665039 on Build GRCh38
UniGene ID:
Hs.513633
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
137994
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ADGRG1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145770.2
NP_001139242.1
8, 9
1305
NM_001145771.2
NP_001139243.1
8, 9
1743
NM_001145772.2
NP_001139244.1
8, 9
1769
NM_001145773.2
NP_001139245.1
7, 8
1640
NM_001145774.2
NP_001139246.1
8, 9
1542
NM_001290142.1
NP_001277071.1
7, 8
845
NM_001290143.1
NP_001277072.1
7, 8
1194
NM_001290144.1
NP_001277073.1
7, 8
1381
NM_005682.6
NP_005673.3
8, 9
1305
NM_201524.3
NP_958932.1
8, 9
1747
NM_201525.3
NP_958933.1
7, 8
1625
XM_005256237.2
XP_005256294.1
7, 8
1341
XM_005256238.2
XP_005256295.1
8, 9
1680
XM_005256239.2
XP_005256296.1
8, 9
1442
XM_005256240.4
XP_005256297.1
7, 8
1163
XM_005256241.4
XP_005256298.1
7, 8
1167
XM_005256242.4
XP_005256299.1
7, 8
1224
XM_005256244.3
XP_005256301.1
7, 8
1316
XM_005256245.2
XP_005256302.1
7, 8
1367
XM_005256246.2
XP_005256303.1
8, 9
1427
XM_005256247.2
XP_005256304.1
7, 8
1326
XM_005256248.2
XP_005256305.1
7, 8
1391
XM_005256249.3
XP_005256306.1
7, 8
1182
XM_005256251.4
XP_005256308.1
7, 8
1208
XM_005256252.2
XP_005256309.1
7, 8
1152
XM_005256254.2
XP_005256311.1
7, 8
1352
XM_005256255.2
XP_005256312.1
8, 9
1665
XM_006721338.2
XP_006721401.1
8, 9
1463
XM_006721339.3
XP_006721402.1
8, 9
1270
XM_006721340.3
XP_006721403.1
8, 9
1528
XM_006721341.2
XP_006721404.1
8, 9
1485
XM_006721342.2
XP_006721405.1
8, 9
1448
XM_006721343.3
XP_006721406.1
8, 9
1513
XM_006721344.2
XP_006721407.1
8, 9
1470
XM_006721345.3
XP_006721408.1
8, 9
1239
XM_006721346.2
XP_006721409.1
7, 8
1094
XM_006721347.2
XP_006721410.1
8, 9
1680
XM_011523461.2
XP_011521763.1
8, 9
1431
XM_011523462.2
XP_011521764.1
7, 8
1246
XM_011523463.2
XP_011521765.1
8, 9
1256
XM_011523464.2
XP_011521766.1
8, 9
1416
XM_011523465.2
XP_011521767.1
8, 9
1354
XM_011523466.2
XP_011521768.1
7, 8
1401
XM_011523467.2
XP_011521769.1
8, 9
1148
XM_011523468.2
XP_011521770.1
7, 8
1150
XM_017023892.1
XP_016879381.1
7, 8
1352
GenBank mRNA
AF106858.1
-
7, 8
1175
AJ011001.1
-
8, 9
1329
AK075270.1
-
8, 9
1257
AK075555.1
-
8, 9
1280
AK131550.1
-
7, 8
800
AK297236.1
-
7, 8
1254
AK299110.1
-
7, 8
1169
AK312405.1
-
7, 8
1461
AY358400.1
-
8, 9
1226
BC008770.2
-
7, 8
1171
3139174
BC013207.1
-
8, 9
1319
BT007311.1
-
6, 7
1013
CR936747.1
-
7, 8
1136
EU432119.1
-
6, 7
1013
KU178472.1
-
6, 7
1013
KU178473.1
-
6, 7
1013
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
cell adhesion
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
Rho protein signal transduction
cell-cell signaling
brain development
negative regulation of cell proliferation
vascular endothelial growth factor production
cell migration
cerebral cortex regionalization
cerebral cortex radial glia guided migration
layer formation in cerebral cortex
positive regulation of Rho protein signal transduction
positive regulation of cell adhesion
protein kinase C signaling
positive regulation of neural precursor cell proliferation
negative regulation of neuron migration
G-protein coupled receptor activity
collagen binding
heparin binding
extracellular matrix binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-97l6p:80/100.66.76.85:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline