Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FN3KRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024619.3 | NP_078895.2 | 6 | 1620 | ||
NR_046408.1 | 7 | 1748 | ||||
GenBank mRNA | AK022233.1 | - | 6 | 1587 | ||
AL136631.1 | - | 6 | 1591 | |||
AY360465.1 | - | 6 | 1587 | |||
BC001458.2 | - | 3 | 1157 | |||
BC007611.1 | - | 6 | 1595 | 3351601 | ||
BC014408.1 | - | 6 | 1574 | 4652087 |