Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005707.1 | NP_005698.1 | 5 | 1902 | ||
GenBank mRNA | AK096970.1 | - | 5 | 1902 | ||
BC016992.2 | - | 3 | 832 | |||
BC092464.1 | - | 7 | 1226 |