Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005735.3 | NP_005726.1 | 3 | 381 | ||
XM_017003116.1 | XP_016858605.1 | 3 | 435 | |||
GenBank mRNA | BC004374.2 | - | 3 | 249 | 3638252 | |
BC006372.2 | - | 3 | 214 | 4138683 | ||
BC010090.2 | - | 3 | 281 | 2989545 | ||
X82207.1 | - | 3 | 229 | |||
X82208.1 | - | 2 | 90 |