Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003886.2 | NP_003877.2 | 2 | 207 | ||
NM_139289.1 | NP_647450.1 | 2 | 180 | |||
GenBank mRNA | AF072756.1 | - | 2 | 235 | ||
AK292463.1 | - | 2 | 201 | |||
BC033007.1 | - | 2 | 204 | |||
BC070150.1 | - | 2 | 199 | |||
BC126250.1 | - | 2 | 206 | 8991966 | ||
BC126252.1 | - | 2 | 206 | 8991968 | ||
Y15195.1 | - | 2 | 207 |