Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005735.3 | NP_005726.1 | 8, 9 | 1141 | ||
XM_005263854.4 | XP_005263911.1 | 7, 8 | 900 | |||
XM_017003116.1 | XP_016858605.1 | 8, 9 | 1195 | |||
GenBank mRNA | BC004374.2 | - | 8, 9 | 1009 | 3638252 | |
BC006372.2 | - | 8, 9 | 974 | 4138683 | ||
BC010090.2 | - | 8, 9 | 1041 | 2989545 | ||
BC025274.2 | - | 3, 4 | 492 | |||
X82207.1 | - | 8, 9 | 989 | |||
X82208.1 | - | 7, 8 | 850 |