Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005762.2 | NP_005753.1 | 14, 15 | 2400 | ||
GenBank mRNA | AK131063.1 | - | 7, 8 | 988 | ||
AK225760.1 | - | 14, 15 | 2400 | |||
AK313478.1 | - | 14, 15 | 2117 | |||
AL360114.1 | - | 2, 3 | 226 | |||
AL360188.1 | - | 2, 3 | 307 | |||
AL360189.1 | - | 2, 3 | 187 | |||
BC004978.1 | - | 14, 15 | 2400 | 2906325 | ||
BC007390.2 | - | 3, 4 | 717 | |||
BC052986.2 | - | 12, 13 | 2151 | 6502540 | ||
U31657.1 | - | 3, 4 | 656 | |||
U78773.1 | - | 14, 15 | 2211 | |||
U95040.1 | - | 14, 15 | 2367 | |||
X97548.1 | - | 14, 15 | 2472 |