Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHFPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005779.2 | NP_005770.1 | 5 | 2173 | ||
XM_006714515.1 | XP_006714578.1 | 4 | 1860 | |||
GenBank mRNA | AK125227.1 | - | 2 | 1670 | ||
AY309920.1 | - | 2 | 1572 | |||
BC136335.1 | - | 2 | 1524 | 9020322 | ||
BC136336.1 | - | 2 | 1524 | 9020323 | ||
CR936660.1 | - | 3 | 1810 | |||
D86961.1 | - | 2 | 1392 |