Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHFP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005780.2 | NP_005771.1 | 4 | 1107 | ||
XM_011534861.1 | XP_011533163.1 | 4 | 910 | |||
GenBank mRNA | AF098807.1 | - | 4 | 952 | ||
AK129819.1 | - | 4 | 608 | |||
AK223369.1 | - | 4 | 608 | |||
BC017824.1 | - | 4 | 955 | 4696562 |