Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005783.3 | NP_005774.2 | 5 | 1423 | ||
GenBank mRNA | AB006679.1 | - | 5 | 1360 | ||
BC005968.1 | - | 5 | 1359 | 4073821 | ||
BC070183.1 | - | 5 | 1357 | 4551680 |