Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242826.1 | NP_001229755.1 | 7 | 1872 | ||
NM_005785.3 | NP_005776.1 | 7 | 1933 | |||
NM_194358.2 | NP_919339.1 | 8 | 2076 | |||
NM_194359.2 | NP_919340.1 | 7 | 1828 | |||
NR_040053.1 | 9 | 2219 | ||||
XM_005268561.4 | XP_005268618.1 | 8 | 1984 | |||
XM_011537734.2 | XP_011536036.1 | 8 | 1990 | |||
XM_011537735.2 | XP_011536037.1 | 8 | 1907 | |||
GenBank mRNA | AF077599.1 | - | 6 | 1766 | ||
AL080105.1 | - | 1 | 759 | |||
AY007109.1 | - | 2 | 1051 | |||
BC024284.1 | - | 8 | 2054 | |||
BC032637.1 | - | 6 | 1781 | 5496849 | ||
BC036267.1 | - | 7 | 1877 | |||
BC095418.1 | - | 9 | 2154 |