Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSHZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308210.1 | NP_001295139.1 | 2 | 2416 | ||
NM_005786.5 | NP_005777.3 | 2 | 2517 | |||
XM_005266641.3 | XP_005266698.1 | 2 | 2787 | |||
GenBank mRNA | AB210042.1 | - | 1 | 4612 | ||
AF039698.1 | - | 1 | 1315 | |||
AK127042.1 | - | 1 | 3258 | |||
AK223545.1 | - | 1 | 1113 | |||
AL833321.1 | - | 2 | 2085 | |||
BC024152.2 | - | 1 | 1004 |