Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005811.3 | NP_005802.1 | 2 | 755 | ||
XM_006719194.3 | XP_006719257.1 | 2 | 757 | |||
GenBank mRNA | AF028333.1 | - | 2 | 636 | ||
AF100907.1 | - | 2 | 756 | |||
KC561773.1 | - | 1 | 377 |