Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078645.1 | NP_001072113.1 | 3 | 320 | ||
NM_001318517.1 | NP_001305446.1 | 3 | 317 | |||
NM_001318518.1 | NP_001305447.1 | 3 | 317 | |||
NM_003903.3 | NP_003894.3 | 3 | 320 | |||
XM_005266206.1 | XP_005266263.1 | 3 | 362 | |||
XM_006719992.1 | XP_006720055.1 | 3 | 307 | |||
XM_006719993.1 | XP_006720056.1 | 2 | 255 | |||
XM_011534851.1 | XP_011533153.1 | 3 | 362 | |||
XM_011534853.1 | XP_011533155.1 | 3 | 362 | |||
XM_017020830.1 | XP_016876319.1 | 3 | 362 | |||
XM_017020831.1 | XP_016876320.1 | 3 | 362 | |||
XM_017020832.1 | XP_016876321.1 | 2 | 255 | |||
GenBank mRNA | AB209850.1 | - | 2 | 1473 | ||
AF164598.1 | - | 3 | 123 | |||
AK296424.1 | - | 3 | 310 | |||
AK315302.1 | - | 3 | 151 | |||
AL540490.3 | - | 3 | 316 | |||
BC010875.2 | - | 3 | 224 | 3879150 | ||
BC017244.1 | - | 3 | 230 | 4877392 | ||
DC323610.1 | - | 3 | 323 | |||
U18291.1 | - | 3 | 143 |