Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAX1BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079864.2 | NP_001073333.1 | 4 | 514 | ||
NM_001206901.1 | NP_001193830.1 | 4 | 517 | |||
NM_006024.6 | NP_006015.4 | 4 | 514 | |||
XM_005249900.2 | XP_005249957.1 | 4 | 514 | |||
XM_017012754.1 | XP_016868243.1 | 4 | 514 | |||
GenBank mRNA | AF090891.1 | - | 4 | 432 | ||
AF268075.1 | - | 3 | 332 | |||
AK000098.1 | - | 4 | 427 | |||
AK299729.1 | - | 4 | 517 | |||
AK301064.1 | - | 4 | 453 | |||
AK314292.1 | - | 4 | 484 | |||
AL136586.1 | - | 4 | 420 | |||
BC050358.2 | - | 4 | 399 | 6055189 | ||
BP280714.1 | - | 4 | 490 | |||
CR457056.1 | - | 3 | 332 | |||
CR533556.1 | - | 3 | 332 | |||
DC424592.1 | - | 4 | 512 | |||
U33821.2 | - | 4 | 415 |