Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNMA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006029.4 | NP_006020.4 | 1 | 1997 | ||
GenBank mRNA | AF320308.1 | - | 1 | 1482 | ||
AK290980.1 | - | 1 | 1334 | |||
AL834327.1 | - | 1 | 730 | |||
BC039577.1 | - | 1 | 1300 | 4431875 |