Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003909.4 | NP_003900.1 | 5 | 541 | ||
XM_005251093.4 | XP_005251150.1 | 4 | 493 | |||
XM_017013945.1 | XP_016869434.1 | 6 | 666 | |||
GenBank mRNA | AB014536.1 | - | 5 | 498 | ||
AF077226.2 | - | 4 | 439 | |||
AK292912.1 | - | 5 | 530 | |||
BC007017.1 | - | 5 | 525 | |||
BC008385.1 | - | 5 | 525 | |||
BC036242.1 | - | 5 | 501 | 5300530 | ||
BC066597.1 | - | 5 | 512 | 6168840 | ||
DA780310.1 | - | 5 | 542 | |||
DB135485.1 | - | 5 | 541 |