Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272071.1 | NP_001259000.1 | 3 | 548 | ||
NM_003916.4 | NP_003907.3 | 3 | 548 | |||
XM_005274614.3 | XP_005274671.1 | 3 | 500 | |||
XM_011545599.1 | XP_011543901.1 | 3 | 500 | |||
XM_017029925.1 | XP_016885414.1 | 3 | 500 | |||
XM_017029926.1 | XP_016885415.1 | 3 | 435 | |||
GenBank mRNA | AB015320.1 | - | 2 | 221 | ||
AF087876.1 | - | 2 | 339 | |||
AF251295.1 | - | 3 | 301 | |||
AK130075.1 | - | 3 | 350 | |||
AK296135.1 | - | 3 | 347 | |||
AK299921.1 | - | 3 | 347 | |||
AK302908.1 | - | 3 | 318 | |||
AK312594.1 | - | 3 | 347 | |||
BC001117.2 | - | 3 | 295 | 3050001 | ||
BC071867.1 | - | 3 | 297 | 4556064 | ||
BI462192.1 | - | 3 | 513 | |||
BT006738.1 | - | 2 | 221 | |||
BX537780.1 | - | 2 | 149 |
Gene Symbol: LOC653653 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027408.1 | 1 | 478 | |||
GenBank mRNA | AK295685.1 | - | 1 | 478 |