Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185072.2 | NP_001172001.1 | 4 | 1188 | ||
NM_001185073.2 | NP_001172002.1 | 3 | 1149 | |||
NM_012129.4 | NP_036261.1 | 3 | 1145 | |||
GenBank mRNA | AL136770.1 | - | 3 | 997 | ||
BC036754.2 | - | 2 | 934 | 5201423 | ||
BC068532.1 | - | 3 | 1031 | 5294796 |