Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136554.1 | NP_001130026.1 | 3 | 338 | ||
NM_012290.4 | NP_036422.3 | 3 | 779 | |||
XM_011512238.2 | XP_011510540.1 | 3 | 819 | |||
XM_017005421.1 | XP_016860910.1 | 3 | 819 | |||
XM_017005422.1 | XP_016860911.1 | 2 | 700 | |||
GenBank mRNA | AF162666.1 | - | 3 | 460 | ||
AK090779.1 | - | 3 | 338 | |||
AK091975.1 | - | 3 | 779 | |||
AK292508.1 | - | 3 | 420 | |||
BC032657.1 | - | 3 | 340 | 5532877 | ||
D50927.2 | - | 3 | 412 | |||
DA144429.1 | - | 3 | 338 |