Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135662.1 | NP_001129134.1 | 2 | 334 | ||
NM_001135663.1 | NP_001129135.1 | 1 | 621 | |||
NM_003929.2 | NP_003920.1 | 2 | 435 | |||
XM_005245569.1 | XP_005245626.1 | 2 | 333 | |||
XM_005245570.1 | XP_005245627.1 | 2 | 386 | |||
XM_005245571.1 | XP_005245628.1 | 2 | 311 | |||
XM_017002748.1 | XP_016858237.1 | 2 | 408 | |||
XM_017002749.1 | XP_016858238.1 | 2 | 386 | |||
XM_017002750.1 | XP_016858239.1 | 2 | 318 | |||
GenBank mRNA | AK223514.1 | - | 1 | 211 | ||
AK303879.1 | - | 1 | 372 | |||
AK311085.1 | - | 2 | 312 | |||
BC002585.2 | - | 2 | 268 | 3140744 | ||
BT019540.1 | - | 1 | 95 | |||
BX647960.1 | - | 2 | 400 | |||
CR542010.1 | - | 1 | 95 | |||
D84488.1 | - | 1 | 135 | |||
DA284582.1 | - | 2 | 334 | |||
DC365059.1 | - | 2 | 419 |