Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ULK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142610.1 | NP_001136082.1 | 1, 2 | 601 | ||
NM_014683.3 | NP_055498.3 | 1, 2 | 601 | |||
XM_017025424.1 | XP_016880913.1 | 1, 2 | 426 | |||
XM_017025425.1 | XP_016880914.1 | 1, 2 | 426 | |||
XM_017025426.1 | XP_016880915.1 | 1, 2 | 426 | |||
GenBank mRNA | AB014523.1 | - | 1, 2 | 592 | ||
AK314345.1 | - | 1, 2 | 126 | |||
BC034988.1 | - | 2, 3 | 166 | 5268025 |