Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SART3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014706.3 | NP_055521.1 | 6 | 1068 | ||
XM_005269241.4 | XP_005269298.1 | 6 | 1135 | |||
XM_011539026.2 | XP_011537328.1 | 5 | 521 | |||
GenBank mRNA | AB020880.1 | - | 6 | 852 | ||
AF387506.1 | - | 6 | 847 | |||
AK225174.1 | - | 6 | 847 | |||
AK290209.1 | - | 6 | 860 | |||
AK299250.1 | - | 5 | 559 | |||
AK304834.1 | - | 7 | 770 | |||
BC032601.1 | - | 6 | 851 | |||
BC093784.1 | - | 6 | 838 | 7939629 | ||
BC103706.1 | - | 6 | 836 | |||
BC111983.1 | - | 6 | 838 | 8327451 | ||
BC143253.1 | - | 6 | 838 | 9051755 | ||
CR933631.1 | - | 1 | 1203 | |||
D63879.1 | - | 6 | 833 |