Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KNTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014708.4 | NP_055523.1 | 5, 6 | 616 | ||
XM_006719706.2 | XP_006719769.1 | 5, 6 | 598 | |||
XM_011539027.2 | XP_011537329.1 | 5, 6 | 598 | |||
XM_011539029.2 | XP_011537331.1 | 5, 6 | 598 | |||
XM_011539030.1 | XP_011537332.1 | 5, 6 | 598 | |||
XM_017020254.1 | XP_016875743.1 | 5, 6 | 598 | |||
XM_017020255.1 | XP_016875744.1 | 4, 5 | 562 | |||
XM_017020257.1 | XP_016875746.1 | 5, 6 | 598 | |||
GenBank mRNA | BC150278.1 | - | 5, 6 | 616 | 9007259 | |
D79988.1 | - | 5, 6 | 616 |