Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142503.2 | NP_001135975.1 | 9 | 2316 | ||
NM_001142504.2 | NP_001135976.1 | 8 | 2210 | |||
NM_014725.4 | NP_055540.2 | 8 | 2234 | |||
XM_005262314.4 | XP_005262371.1 | 9 | 2302 | |||
XM_005262315.3 | XP_005262372.1 | 8 | 2222 | |||
XM_011531069.2 | XP_011529371.1 | 8 | 2090 | |||
XM_011531070.2 | XP_011529372.1 | 8 | 2103 | |||
GenBank mRNA | AK291747.1 | - | 8 | 2210 | ||
BC035587.1 | - | 8 | 2245 | 5518429 | ||
CR749411.1 | - | 9 | 2217 | |||
D80011.1 | - | 8 | 2199 |