Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASSF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014737.2 | NP_055552.1 | 11, 12 | 1104 | ||
NM_170774.1 | NP_739580.1 | 10, 11 | 962 | |||
XM_005260895.3 | XP_005260952.1 | 11, 12 | 1170 | |||
XM_011529410.1 | XP_011527712.1 | 11, 12 | 1104 | |||
XM_011529411.1 | XP_011527713.1 | 11, 12 | 1176 | |||
XM_011529412.2 | XP_011527714.1 | 10, 11 | 1025 | |||
XM_017028149.1 | XP_016883638.1 | 10, 11 | 999 | |||
XM_017028150.1 | XP_016883639.1 | 12, 13 | 1307 | |||
XM_017028151.1 | XP_016883640.1 | 12, 13 | 1274 | |||
XM_017028152.1 | XP_016883641.1 | 12, 13 | 1351 | |||
XM_017028153.1 | XP_016883642.1 | 10, 11 | 1052 | |||
GenBank mRNA | AK222650.1 | - | 11, 12 | 1067 | ||
AK291458.1 | - | 11, 12 | 1059 | |||
AY154470.1 | - | 11, 12 | 1104 | |||
AY154471.1 | - | 8, 9 | 878 | |||
AY154472.1 | - | 10, 11 | 962 | |||
BC110385.1 | - | 8, 9 | 878 | |||
BC117118.1 | - | 9, 10 | 941 | 40125669 | ||
BC117120.1 | - | 9, 10 | 941 | 40125671 | ||
CR627436.1 | - | 10, 11 | 4311 | |||
D79990.1 | - | 11, 12 | 1104 |