Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASSF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014737.2 | NP_055552.1 | 12 | 4174 | ||
NM_170774.1 | NP_739580.1 | 11 | 4032 | |||
XM_005260895.3 | XP_005260952.1 | 12 | 4240 | |||
XM_011529410.1 | XP_011527712.1 | 12 | 4174 | |||
XM_011529411.1 | XP_011527713.1 | 12 | 4246 | |||
XM_011529412.2 | XP_011527714.1 | 11 | 4095 | |||
XM_017028149.1 | XP_016883638.1 | 11 | 4069 | |||
XM_017028150.1 | XP_016883639.1 | 13 | 4377 | |||
XM_017028151.1 | XP_016883640.1 | 13 | 4344 | |||
XM_017028152.1 | XP_016883641.1 | 13 | 4421 | |||
XM_017028153.1 | XP_016883642.1 | 11 | 4122 | |||
GenBank mRNA | AY154470.1 | - | 12 | 4174 | ||
AY154471.1 | - | 9 | 3948 | |||
AY154472.1 | - | 11 | 4032 | |||
CR627436.1 | - | 11 | 7382 | |||
D79990.1 | - | 12 | 4174 |