Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014740.3 | NP_055555.1 | 3, 4 | 539 | ||
GenBank mRNA | AK290608.1 | - | 3, 4 | 520 | ||
BC003662.2 | - | 3, 4 | 449 | 2988931 | ||
BC004386.1 | - | 3, 4 | 470 | 3619386 | ||
BC011151.1 | - | 3, 4 | 474 | 3452439 | ||
CR456750.1 | - | 3, 4 | 317 | |||
D21853.1 | - | 3, 4 | 531 | |||
DA667983.1 | - | 3, 4 | 539 | |||
X79538.1 | - | 3, 4 | 390 |