Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM9SF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014742.3 | NP_055557.2 | 3 | 438 | ||
XM_005260622.4 | XP_005260679.1 | 3 | 490 | |||
XM_017028154.1 | XP_016883643.1 | 3 | 219 | |||
XM_017028155.1 | XP_016883644.1 | 3 | 490 | |||
XM_017028156.1 | XP_016883645.1 | 3 | 230 | |||
XM_017028157.1 | XP_016883646.1 | 3 | 487 | |||
XM_017028158.1 | XP_016883647.1 | 3 | 225 | |||
XM_017028159.1 | XP_016883648.1 | 3 | 271 | |||
XM_017028160.1 | XP_016883649.1 | 3 | 227 | |||
XM_017028161.1 | XP_016883650.1 | 3 | 219 | |||
GenBank mRNA | AK126534.1 | - | 3 | 230 | ||
AK294980.1 | - | 3 | 230 | |||
AK296500.1 | - | 3 | 218 | |||
AK302439.1 | - | 3 | 216 | |||
AK316293.1 | - | 2 | 142 | |||
BC021107.1 | - | 3 | 214 | 4896167 | ||
BC022850.2 | - | 2 | 196 | |||
D87444.1 | - | 3 | 492 | |||
DC381660.1 | - | 3 | 438 |