Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM9SF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014742.3 | NP_055557.2 | 7 | 904 | ||
XM_005260622.4 | XP_005260679.1 | 7 | 956 | |||
XM_017028154.1 | XP_016883643.1 | 7 | 685 | |||
XM_017028155.1 | XP_016883644.1 | 7 | 956 | |||
XM_017028156.1 | XP_016883645.1 | 7 | 652 | |||
XM_017028157.1 | XP_016883646.1 | 7 | 909 | |||
XM_017028158.1 | XP_016883647.1 | 7 | 647 | |||
XM_017028159.1 | XP_016883648.1 | 7 | 709 | |||
XM_017028160.1 | XP_016883649.1 | 7 | 621 | |||
XM_017028161.1 | XP_016883650.1 | 7 | 657 | |||
GenBank mRNA | AK126534.1 | - | 5 | 354 | ||
AK294980.1 | - | 7 | 652 | |||
AK296500.1 | - | 7 | 561 | |||
AK302439.1 | - | 8 | 759 | |||
AK316293.1 | - | 6 | 608 | |||
BC021107.1 | - | 7 | 680 | 4896167 | ||
BC022850.2 | - | 6 | 662 | |||
D87444.1 | - | 7 | 958 |