Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIEZO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142864.3 | NP_001136336.2 | 18 | 2697 | ||
GenBank mRNA | AB161230.1 | - | 7 | 1148 | ||
BC150271.1 | - | 7 | 985 | 9007243 | ||
D87071.1 | - | 7 | 985 | |||
KC602455.1 | - | 18 | 2441 |