Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIEZO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142864.3 | NP_001136336.2 | 26 | 4028 | ||
GenBank mRNA | AB161230.1 | - | 15 | 2479 | ||
BC150271.1 | - | 15 | 2316 | 9007243 | ||
D87071.1 | - | 15 | 2316 | |||
KC602455.1 | - | 26 | 3772 |