Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267059.1 | NP_001253988.1 | 4 | 501 | ||
NM_001267060.1 | NP_001253989.1 | 3 | 462 | |||
NM_001267061.1 | NP_001253990.1 | 4 | 372 | |||
NM_014748.3 | NP_055563.1 | 4 | 537 | |||
NR_049782.1 | 4 | 662 | ||||
NR_049783.1 | 5 | 635 | ||||
NR_049784.1 | 5 | 611 | ||||
NR_049785.1 | 3 | 544 | ||||
NR_049786.1 | 4 | 493 | ||||
NR_049787.1 | 2 | 344 | ||||
NR_049788.1 | 2 | 344 | ||||
XM_011533203.1 | XP_011531505.1 | 3 | 311 | |||
XM_017005405.1 | XP_016860894.1 | 3 | 311 | |||
GenBank mRNA | AK129689.1 | - | 4 | 421 | ||
AK222543.1 | - | 4 | 420 | |||
AK293252.1 | - | 5 | 511 | |||
AK293464.1 | - | 4 | 353 | |||
AK295278.1 | - | 2 | 223 | |||
AK297054.1 | - | 4 | 546 | |||
AK298620.1 | - | 4 | 372 | |||
AK298856.1 | - | 4 | 374 | |||
AK298869.1 | - | 3 | 323 | |||
AK300144.1 | - | 4 | 384 | |||
AK300641.1 | - | 2 | 217 | |||
AK307122.1 | - | 3 | 420 | |||
AK308487.1 | - | 3 | 423 | |||
AK309640.1 | - | 3 | 295 | |||
AK316373.1 | - | 4 | 316 | |||
BC002524.1 | - | 4 | 398 | 3140610 | ||
BC002610.1 | - | 4 | 387 | 3143104 | ||
BC014620.2 | - | 4 | 398 | 3010648 | ||
BC050590.1 | - | 4 | 392 | 6044270 | ||
BT007167.1 | - | 4 | 289 | |||
CR457081.1 | - | 4 | 289 | |||
D31764.1 | - | 4 | 511 | |||
DB177194.1 | - | 4 | 527 |