Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLGAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146015.1 | NP_001139487.1 | 3 | 568 | ||
NM_014750.4 | NP_055565.3 | 3 | 568 | |||
XM_017021840.1 | XP_016877329.1 | 3 | 460 | |||
GenBank mRNA | AB076695.1 | - | 3 | 568 | ||
AK291847.1 | - | 3 | 445 | |||
AK299338.1 | - | 3 | 445 | |||
BC010658.1 | - | 3 | 460 | 3873367 | ||
BC016276.2 | - | 2 | 354 | 3537754 | ||
BM788630.1 | - | 3 | 510 | |||
BT007344.1 | - | 1 | 108 | |||
BX248255.1 | - | 3 | 434 | |||
D13633.3 | - | 3 | 441 |