Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MTSS1 SIRNA Assays ›
Gene Symbol:
MTSS1
Gene Name:
metastasis suppressor 1
Gene Aliases:
MIM, MIMA, MIMB
Chromosome Location:
Chr.8: 124550770 - 124728507 on Build GRCh38
UniGene ID:
Hs.336994
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
138404
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MTSS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282971.1
NP_001269900.1
15
4326
NM_001282974.1
NP_001269903.1
14
4239
NM_014751.5
NP_055566.3
14
4314
XM_005251111.1
XP_005251168.1
14
4619
XM_005251113.1
XP_005251170.1
13
4424
XM_005251118.1
XP_005251175.1
14
4097
XM_006716700.1
XP_006716763.1
14
4616
XM_006716701.1
XP_006716764.1
13
4607
XM_006716702.1
XP_006716765.1
14
4568
XM_006716703.1
XP_006716766.1
13
4373
XM_006716704.1
XP_006716767.1
15
4289
XM_006716705.1
XP_006716768.1
15
4241
XM_006716706.1
XP_006716769.1
14
4046
XM_011517403.1
XP_011515705.1
13
4073
XM_011517404.2
XP_011515706.1
12
15785
XM_011517407.1
XP_011515709.1
5
3273
XM_017014086.1
XP_016869575.1
13
4604
XM_017014087.1
XP_016869576.1
14
4544
XM_017014088.1
XP_016869577.1
13
4532
XM_017014089.1
XP_016869578.1
13
4529
XM_017014090.1
XP_016869579.1
12
5165
XM_017014091.1
XP_016869580.1
14
4053
XM_017014092.1
XP_016869581.1
12
4412
XM_017014093.1
XP_016869582.1
14
4277
XM_017014094.1
XP_016869583.1
14
4229
XM_017014095.1
XP_016869584.1
15
4217
XM_017014096.1
XP_016869585.1
15
4214
XM_017014097.1
XP_016869586.1
14
4202
XM_017014098.1
XP_016869587.1
13
4085
XM_017014099.1
XP_016869588.1
13
4082
XM_017014100.1
XP_016869589.1
13
4034
XM_017014101.1
XP_016869590.1
12
3864
GenBank mRNA
AB007889.2
-
11
3496
AF086645.1
-
14
4224
AK126651.1
-
2
3024
AK126989.1
-
7
3070
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of defense response to virus by host
movement of cell or subcellular component
plasma membrane organization
cell adhesion
transmembrane receptor protein tyrosine kinase signaling pathway
microspike assembly
actin cytoskeleton organization
negative regulation of epithelial cell proliferation
renal tubule morphogenesis
cellular response to fluid shear stress
glomerulus morphogenesis
nephron tubule epithelial cell differentiation
xenophagy
epithelial cell proliferation involved in renal tubule morphogenesis
actin monomer binding
receptor binding
protein binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-slhn9:80/100.66.78.111:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline