Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GS1-124K5.11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111972.1 | 5 | 1463 | |||
NR_111974.1 | 3 | 1188 | ||||
NR_111975.1 | 4 | 1285 | ||||
NR_111976.1 | 4 | 1366 | ||||
NR_111977.1 | 6 | 1532 | ||||
NR_111978.1 | 5 | 1460 | ||||
NR_111979.1 | 4 | 1363 | ||||
GenBank mRNA | AK054923.1 | - | 4 | 1953 | ||
AK126730.1 | - | 4 | 2891 | |||
BC103766.1 | - | 5 | 1432 |
Gene Symbol: TOMM20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014765.2 | NP_055580.1 | 4 | 498 | ||
GenBank mRNA | AK289810.1 | - | 4 | 498 | ||
BC000882.1 | - | 4 | 390 | 3462090 | ||
BC066335.1 | - | 4 | 386 | 4818590 | ||
BC071994.1 | - | 4 | 369 | 6082526 | ||
BC100286.1 | - | 4 | 380 | 6157036 | ||
BC107851.1 | - | 4 | 376 | 5579754 | ||
CR456718.1 | - | 4 | 272 | |||
D13641.1 | - | 4 | 373 |