Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207033.1 | NP_001193962.1 | 11 | 2148 | ||
NM_001207034.1 | NP_001193963.1 | 9 | 1848 | |||
NM_001286572.2 | NP_001273501.1 | 11 | 1509 | |||
NM_014771.3 | NP_055586.1 | 11 | 2148 | |||
XM_011545997.1 | XP_011544299.1 | 11 | 1489 | |||
GenBank mRNA | AB014561.1 | - | 11 | 1420 | ||
AF122819.1 | - | 11 | 1424 | |||
AK027406.1 | - | 9 | 1138 | |||
AK291666.1 | - | 11 | 1438 | |||
AK297531.1 | - | 5 | 554 | |||
BC006133.1 | - | 11 | 1415 | 3628183 | ||
BC011769.2 | - | 11 | 1415 | 3607432 | ||
BC018647.2 | - | 11 | 1415 | 2967275 | ||
BC030802.1 | - | 3 | 309 | |||
BX640763.1 | - | 8 | 782 | |||
CR627431.1 | - | 11 | 1477 | |||
KU178492.1 | - | 10 | 1328 | |||
KU178493.1 | - | 10 | 1328 |