Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127460.2 | NP_001120932.1 | 3 | 793 | ||
NM_001166279.1 | NP_001159751.1 | 3 | 722 | |||
NM_014871.4 | NP_055686.3 | 3 | 722 | |||
GenBank mRNA | AB014610.2 | - | 3 | 619 | ||
AB107585.1 | - | 2 | 416 | |||
AK310822.1 | - | 3 | 606 | |||
BC024043.1 | - | 3 | 635 | 3357890 | ||
BC094885.1 | - | 3 | 790 | 4823602 | ||
BX648106.1 | - | 3 | 710 |