Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEM1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015322.4 | NP_056137.1 | 1 | 701 | ||
GenBank mRNA | AB007856.2 | - | 1 | 86 | ||
AF178632.1 | - | 1 | 86 | |||
AF204883.1 | - | 1 | 86 | |||
AK290167.1 | - | 1 | 435 | |||
BC010122.2 | - | 1 | 701 | 3840453 | ||
BC014558.2 | - | 1 | 701 | 3840377 |