Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM4SF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003963.2 | NP_003954.2 | 4, 5 | 608 | ||
GenBank mRNA | AF027204.1 | - | 4, 5 | 609 | ||
BC093688.1 | - | 4, 5 | 605 | 7939533 | ||
BC117163.1 | - | 4, 5 | 605 | 40125714 | ||
CR456925.1 | - | 577 |