Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMMECR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025580.1 | NP_001020751.1 | 4 | 776 | ||
NM_001171689.1 | NP_001165160.1 | 7 | 1074 | |||
NM_015365.2 | NP_056180.1 | 5 | 887 | |||
GenBank mRNA | AJ007014.1 | - | 5 | 974 | ||
AK001703.1 | - | 5 | 410 | |||
AK091430.1 | - | 7 | 1074 | |||
AK314596.1 | - | 5 | 818 | |||
BC060813.1 | - | 4 | 709 | 30344824 | ||
BX648797.1 | - | 4 | 738 | |||
KU178496.1 | - | 4 | 695 | |||
KU178497.1 | - | 5 | 806 |