Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166588.2 | NP_001160060.1 | 5 | 709 | ||
NM_001261418.1 | NP_001248347.1 | 5 | 709 | |||
NM_020365.4 | NP_065098.1 | 5 | 709 | |||
XM_017002745.1 | XP_016858234.1 | 5 | 597 | |||
XM_017002746.1 | XP_016858235.1 | 6 | 676 | |||
XM_017002747.1 | XP_016858236.1 | 6 | 670 | |||
GenBank mRNA | AF257077.1 | - | 5 | 572 | ||
AK022399.1 | - | 5 | 571 | |||
AK024006.1 | - | 5 | 597 | |||
AK314668.1 | - | 5 | 582 | |||
AL834288.1 | - | 5 | 574 | |||
BC018728.1 | - | 5 | 567 | 4849789 | ||
BC021017.1 | - | 5 | 560 |