Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003970.3 | NP_003961.3 | 3, 4 | 395 | ||
XM_006716237.1 | XP_006716300.1 | 3, 4 | 360 | |||
GenBank mRNA | AL596730.1 | - | 3, 4 | 421 | ||
BC052969.1 | - | 3, 4 | 348 | 6205359 | ||
X69089.1 | - | 3, 4 | 305 |