Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021090.3 | NP_066576.1 | 6, 7 | 629 | ||
NM_153050.2 | NP_694690.1 | 6, 7 | 629 | |||
NM_153051.2 | NP_694691.1 | 6, 7 | 629 | |||
XM_005261803.2 | XP_005261860.1 | 7, 8 | 747 | |||
XM_005261804.3 | XP_005261861.1 | 6, 7 | 664 | |||
XM_005261805.2 | XP_005261862.1 | 7, 8 | 747 | |||
XM_005261806.2 | XP_005261863.1 | 6, 7 | 629 | |||
XM_005261808.2 | XP_005261865.1 | 7, 8 | 747 | |||
XM_017029028.1 | XP_016884517.1 | 5, 6 | 521 | |||
XM_017029029.1 | XP_016884518.1 | 5, 6 | 511 | |||
XM_017029030.1 | XP_016884519.1 | 5, 6 | 527 | |||
XM_017029031.1 | XP_016884520.1 | 5, 6 | 515 | |||
XM_017029032.1 | XP_016884521.1 | 6, 7 | 512 | |||
XM_017029034.1 | XP_016884523.1 | 6, 7 | 602 | |||
GenBank mRNA | AB002369.1 | - | 6, 7 | 548 | ||
AF233436.2 | - | 3, 4 | 310 | |||
AF233437.1 | - | 3, 4 | 310 | |||
AF233438.1 | - | 3, 4 | 310 | |||
AK289581.1 | - | 6, 7 | 629 | |||
BC142713.1 | - | 6, 7 | 585 | 40148839 | ||
BC148216.1 | - | 5, 6 | 448 | 40082266 | ||
BC151217.1 | - | 6, 7 | 643 | 40148098 | ||
BC152455.1 | - | 6, 7 | 548 | 8862588 | ||
CR456525.1 | - | 4, 5 | 325 |