Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021999.4 | NP_068839.1 | 6 | 1353 | ||
GenBank mRNA | AF086918.1 | - | 6 | 1245 | ||
AF136973.1 | - | 6 | 1342 | |||
AF152462.1 | - | 6 | 1300 | |||
AF246221.1 | - | 6 | 1310 | |||
AK000503.1 | - | 6 | 1285 | |||
AK027411.1 | - | 6 | 1284 | |||
AK130048.1 | - | 6 | 1289 | |||
BC016148.1 | - | 6 | 1300 | 3912066 |