Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR1I2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003889.3 | NP_003880.3 | 9 | 3553 | ||
NM_022002.2 | NP_071285.1 | 9 | 1879 | |||
NM_033013.2 | NP_148934.1 | 9 | 3442 | |||
GenBank mRNA | AF061056.1 | - | 9 | 2017 | ||
AF084644.1 | - | 9 | 1890 | |||
AF084645.1 | - | 9 | 1993 | |||
AJ009936.1 | - | 10 | 3554 | |||
AJ009937.1 | - | 10 | 3443 | |||
AK122990.1 | - | 6 | 1846 | |||
BC017304.2 | - | 9 | 1950 | 5087401 |