Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPHOSPH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022782.3 | NP_073619.3 | 7, 8 | 1417 | ||
NR_103517.1 | 8, 9 | 1481 | ||||
XM_011537740.2 | XP_011536042.1 | 7, 8 | 1321 | |||
XM_011537741.2 | XP_011536043.1 | 7, 8 | 1321 | |||
XM_017018673.1 | XP_016874162.1 | 7, 8 | 1415 | |||
XM_017018674.1 | XP_016874163.1 | 7, 8 | 1321 | |||
XM_017018675.1 | XP_016874164.1 | 7, 8 | 1349 | |||
XM_017018676.1 | XP_016874165.1 | 7, 8 | 1321 | |||
XM_017018677.1 | XP_016874166.1 | 7, 8 | 1321 | |||
XM_017018678.1 | XP_016874167.1 | 7, 8 | 1267 | |||
XM_017018679.1 | XP_016874168.1 | 8, 9 | 1354 | |||
XM_017018680.1 | XP_016874169.1 | 6, 7 | 1263 | |||
GenBank mRNA | AK023016.1 | - | 2, 3 | 140 | ||
AK091181.1 | - | 3, 4 | 746 | |||
AK096541.1 | - | 8, 9 | 1354 | |||
AL096751.1 | - | 2, 3 | 203 | |||
BC130440.1 | - | 3, 4 | 699 | 40146471 | ||
BC144016.1 | - | 3, 4 | 699 | |||
BC144017.1 | - | 3, 4 | 676 |